siRNA Details
| Gene Symbol: ZNF419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291743.1 | NP_001278672.1 | 2, 3 | 317 | ||
| NM_024691.3 | NP_078967.3 | 2, 3 | 317 | |||
| GenBank mRNA | BC008297.1 | - | 2, 3 | 263 | 3544044 | |
| BC024259.1 | - | 2, 3 | 263 | 3604215 | ||
| CT002615.1 | - | 2, 3 | 247 | |||

ZNF419
Genomic Map