siRNA Details
| Gene Symbol: ZNF750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024702.2 | NP_078978.2 | 2 | 642 | ||
| GenBank mRNA | AK023903.1 | - | 2 | 611 | ||
| BC109036.1 | - | 2 | 601 | 40028398 | ||
| BC109037.1 | - | 2 | 601 | 40028404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF750| Gene Symbol: ZNF750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024702.2 | NP_078978.2 | 2 | 642 | ||
| GenBank mRNA | AK023903.1 | - | 2 | 611 | ||
| BC109036.1 | - | 2 | 601 | 40028398 | ||
| BC109037.1 | - | 2 | 601 | 40028404 | ||