siRNA Details
| Gene Symbol: C1orf115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024709.4 | NP_078985.3 | 1 | 412 | ||
| GenBank mRNA | AK024208.1 | - | 1 | 333 | ||
| AK056749.1 | - | 2 | 278 | |||
| AK091937.1 | - | 1 | 346 | |||
| AK125403.1 | - | 1 | 853 | |||
| BC036067.1 | - | 1 | 378 | 5274863 | ||
| DA175175.1 | - | 1 | 412 | |||

C1orf115
Genomic Map