siRNA Details
| Gene Symbol: CENPU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024629.3 | NP_078905.2 | 10, 11 | 989 | ||
| XM_005263218.4 | XP_005263275.2 | 10, 11 | 1024 | |||
| GenBank mRNA | AF469667.1 | - | 10, 11 | 992 | ||
| AF516710.1 | - | 11, 12 | 1406 | |||
| AK027121.1 | - | 8, 9 | 799 | |||
| AK292297.1 | - | 10, 11 | 935 | |||
| BC031520.1 | - | 5, 6 | 337 | |||
| BC107744.1 | - | 5, 6 | 434 | |||
| BC131556.1 | - | 10, 11 | 989 | 40108113 | ||
| CR457376.1 | - | 3, 4 | 193 | |||
| DQ907910.1 | - | 10, 11 | 919 | |||

CENPU
Genomic Map