siRNA Details
| Gene Symbol: MORC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_001085354.2 | NP_001078823.1 | 5 | 926 | ||
| NM_024657.4 | NP_078933.3 | 5 | 926 | |||
| XM_005262190.3 | XP_005262247.1 | 4 | 816 | |||
| XM_011531027.2 | XP_011529329.1 | 2 | 214 | |||
| XM_017029844.1 | XP_016885333.1 | 4 | 816 | |||
| GenBank mRNA | AK056235.1 | - | 1 | 138 | ||
| AK300305.1 | - | 4 | 501 | |||
| AY927537.1 | - | LIT3543 AY927537 |
3 | 461 | ||
| EF125209.1 | - | 5 | 651 | |||

MORC4
Genomic Map