siRNA Details
| Gene Symbol: IGFLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024660.2 | NP_078936.1 | 5 | 880 | ||
| GenBank mRNA | AK026226.1 | - | 5 | 881 | ||
| AX775837.1 | - | 5 | 883 | |||
| BC031329.1 | - | 2 | 373 | |||
| BC126481.1 | - | 5 | 841 | 8992197 | ||
| BC126483.1 | - | 5 | 841 | 8992199 | ||
| BC144205.1 | - | 5 | 839 | |||

IGFLR1
Genomic Map