siRNA Details
Gene Symbol: ASB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024701.3 | NP_078977.2 | 6 | 1253 | ||
NR_024581.1 | 5 | 1102 | ||||
NR_037164.1 | 7 | 1334 | ||||
GenBank mRNA | AF403031.1 | - | 6 | 1235 | ||
AK023196.1 | - | 6 | 1236 | |||
AK091935.1 | - | 5 | 1102 | |||
AK292071.1 | - | 6 | 1239 | |||
BC012056.1 | - | 7 | 1311 | 4552643 |
