siRNA Details
| Gene Symbol: ELMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024712.3 | NP_078988.2 | 9 | 980 | ||
| GenBank mRNA | AK023886.1 | - | 8 | 744 | ||
| AK056923.1 | - | 8 | 943 | |||
| AK300976.1 | - | 9 | 994 | |||
| BC034410.1 | - | 6 | 592 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ELMO3| Gene Symbol: ELMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024712.3 | NP_078988.2 | 9 | 980 | ||
| GenBank mRNA | AK023886.1 | - | 8 | 744 | ||
| AK056923.1 | - | 8 | 943 | |||
| AK300976.1 | - | 9 | 994 | |||
| BC034410.1 | - | 6 | 592 | |||