siRNA Details
| Gene Symbol: TXNDC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024715.3 | NP_078991.3 | 2 | 928 | ||
| GenBank mRNA | AK026278.1 | - | 3 | 331 | ||
| AK056512.1 | - | 2 | 963 | |||
| AK225454.1 | - | 3 | 331 | |||
| AK225462.1 | - | 3 | 331 | |||
| AK299578.1 | - | 2 | 221 | |||
| AY358649.1 | - | 2 | 356 | |||
| BC001615.1 | - | 2 | 415 | 3544923 | ||
| BC032568.1 | - | 2 | 329 | 5225890 | ||
| BX538131.1 | - | 2 | 255 | |||

TXNDC15
Genomic Map