siRNA Details
| Gene Symbol: LINC00341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026779.1 | 1 | 2138 | |||
| GenBank mRNA | AK024929.1 | - | 1 | 1736 | ||
| BC008299.1 | - | 1 | 2138 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00341| Gene Symbol: LINC00341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026779.1 | 1 | 2138 | |||
| GenBank mRNA | AK024929.1 | - | 1 | 1736 | ||
| BC008299.1 | - | 1 | 2138 | |||