siRNA Details
| Gene Symbol: STEAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205315.1 | NP_001192244.1 | 6 | 2090 | ||
| NM_001205316.1 | NP_001192245.1 | 4 | 1463 | |||
| NM_024636.3 | NP_078912.2 | 5 | 1991 | |||
| GenBank mRNA | AF423422.1 | - | 6 | 1973 | ||
| AF423423.1 | - | 6 | 1974 | |||
| AK026806.1 | - | 5 | 1972 | |||
| AL832955.1 | - | 3 | 1082 | |||
| AL833044.1 | - | 3 | 896 | |||
| AL833671.1 | - | 2 | 812 | |||
| BC020600.1 | - | 4 | 1438 | 4616389 | ||

STEAP4
Genomic Map