siRNA Details
| Gene Symbol: NUP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198887.2 | NP_942590.1 | 8 | 3615 | ||
| NR_104456.1 | 9 | 1980 | ||||
| XM_005266960.4 | XP_005267017.1 | 8 | 4845 | |||
| XM_005266961.3 | XP_005267018.1 | 9 | 3665 | |||
| XM_005266962.3 | XP_005267019.1 | 8 | 5885 | |||
| XM_011535799.2 | XP_011534101.1 | 8 | 4927 | |||
| GenBank mRNA | AK023349.1 | - | 4 | 1937 | ||
| AK074311.1 | - | 2839 | ||||
| AK095994.1 | - | 9 | 1936 | |||
| BC040121.1 | - | 6 | 3260 | |||
| BC046637.1 | - | 1 | 3363 | |||

NUP43
Genomic Map