siRNA Details
| Gene Symbol: NARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243251.1 | NP_001230180.1 | 8, 9 | 824 | ||
| NM_024678.5 | NP_078954.4 | 8, 9 | 1305 | |||
| XM_011545253.1 | XP_011543555.1 | 8, 9 | 1298 | |||
| XM_017018302.1 | XP_016873791.1 | 9 | 1311 | |||
| XM_017018303.1 | XP_016873792.1 | 9, 10 | 889 | |||
| XM_017018304.1 | XP_016873793.1 | 8, 9 | 817 | |||
| GenBank mRNA | AK027094.1 | - | 6, 7 | 613 | ||
| AK056980.1 | - | 8, 9 | 1222 | |||
| AK225444.1 | - | 6, 7 | 585 | |||
| AK315425.1 | - | 8, 9 | 918 | |||
| BC007800.2 | - | 8, 9 | 1115 | 4128365 | ||

NARS2
Genomic Map