siRNA Details
| Gene Symbol: BBS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024685.3 | NP_078961.3 | 2 | 2547 | ||
| GenBank mRNA | AK027213.1 | - | 1 | 931 | ||
| AK225585.1 | - | 1 | 931 | |||
| BC013795.1 | - | 1 | 1139 | |||
| BC026355.1 | - | 2 | 2532 | 4817227 | ||

BBS10
Genomic Map