siRNA Details
| Gene Symbol: ZNF750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024702.2 | NP_078978.2 | 3 | 2685 | ||
| GenBank mRNA | AK023903.1 | - | 3 | 2654 | ||
| AK301690.1 | - | 4 | 1459 | |||
| BC109036.1 | - | 3 | 2640 | 40028398 | ||
| BC109037.1 | - | 3 | 2644 | 40028404 | ||

ZNF750
Genomic Map