siRNA Details
| Gene Symbol: LRRC31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277127.1 | NP_001264056.1 | 9 | 1857 | ||
| NM_001277128.1 | NP_001264057.1 | 8 | 1671 | |||
| NM_024727.3 | NP_079003.2 | 9 | 1839 | |||
| XM_011513158.2 | XP_011511460.1 | 8 | 1671 | |||
| XM_011513159.2 | XP_011511461.1 | 8 | 1671 | |||
| XM_017007204.1 | XP_016862693.1 | 9 | 1560 | |||
| GenBank mRNA | AK026912.1 | - | 8 | 1668 | ||
| AY358138.1 | - | 9 | 1769 | |||
| EU668335.1 | - | 9 | 1788 | |||

LRRC31
Genomic Map