siRNA Details
| Gene Symbol: MOB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024761.4 | NP_079037.3 | 2 | 584 | ||
| GenBank mRNA | AJ580636.1 | - | 1 | 88 | ||
| AK023266.1 | - | 2 | 513 | |||
| AL832572.2 | - | 2 | 478 | |||
| BC033027.1 | - | 2 | 496 | 5268356 | ||
| CR457307.1 | - | 1 | 88 | |||
| DB266074.1 | - | 2 | 513 | |||

MOB3B
Genomic Map