siRNA Details
| Gene Symbol: ZDHHC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024786.2 | NP_079062.1 | 2, 3 | 785 | ||
| XM_017009872.1 | XP_016865361.1 | 3, 4 | 501 | |||
| GenBank mRNA | AK023215.1 | - | 2, 3 | 785 | ||
| BC032000.1 | - | 2, 3 | 404 | 4750082 | ||
| Gene Symbol: ZDHHC11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010117.1 | XP_016865606.1 | 4, 5 | 1637 | ||

ZDHHC11
Genomic Map