siRNA Details
| Gene Symbol: CLUAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015041.2 | NP_055856.1 | 7 | 716 | ||
| NM_024793.2 | NP_079069.1 | 3 | 174 | |||
| XM_006720867.3 | XP_006720930.1 | 6 | 567 | |||
| XM_017023068.1 | XP_016878557.1 | 7 | 731 | |||
| XM_017023069.1 | XP_016878558.1 | 7 | 731 | |||
| XM_017023070.1 | XP_016878559.1 | 7 | 716 | |||
| XM_017023071.1 | XP_016878560.1 | 7 | 1705 | |||
| XM_017023072.1 | XP_016878561.1 | 7 | 1705 | |||
| XM_017023073.1 | XP_016878562.1 | 6 | 567 | |||
| GenBank mRNA | AB014543.1 | - | 7 | 643 | ||
| AB089691.1 | - | 7 | 698 | |||
| AK023359.1 | - | 3 | 174 | |||
| BC017070.2 | - | 7 | 644 | 3916900 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_029738, AK011846, AK134648, BC024910.

CLUAP1
Genomic Map