siRNA Details
Gene Symbol: LINC00115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024321.1 | 1 | 690 | |||
GenBank mRNA | AK026292.1 | - | 1 | 674 | ||
BC017762.1 | - | 1 | 690 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024321.1 | 1 | 690 | |||
GenBank mRNA | AK026292.1 | - | 1 | 674 | ||
BC017762.1 | - | 1 | 690 |