siRNA Details
| Gene Symbol: FLJ13224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026806.1 | 1 | 1120 | |||
| GenBank mRNA | AK023286.1 | - | 1 | 1120 | ||
| BC118557.1 | - | 1 | 947 | |||
| BC118637.1 | - | 1 | 947 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ13224| Gene Symbol: FLJ13224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026806.1 | 1 | 1120 | |||
| GenBank mRNA | AK023286.1 | - | 1 | 1120 | ||
| BC118557.1 | - | 1 | 947 | |||
| BC118637.1 | - | 1 | 947 | |||