siRNA Details
| Gene Symbol: IRF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006084.4 | NP_006075.3 | 2 | 297 | ||
| GenBank mRNA | AK295467.1 | - | 2 | 190 | ||
| BC035716.2 | - | 1 | 173 | 5588434 | ||
| BI919023.1 | - | 2 | 484 | |||
| M87503.1 | - | 2 | 204 | |||

IRF9
Genomic Map