siRNA Details
| Gene Symbol: ZNF671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321376.1 | NP_001308305.1 | 3 | 298 | ||
| NM_024833.2 | NP_079109.2 | 2 | 263 | |||
| XM_017027314.1 | XP_016882803.1 | 2 | 756 | |||
| GenBank mRNA | AK027159.1 | - | 2 | 251 | ||
| AK091421.1 | - | 3 | 298 | |||
| AK225665.1 | - | 2 | 260 | |||
| BC025728.1 | - | 2 | 210 | 5202908 | ||
| DB104548.1 | - | 2 | 263 | |||

ZNF671
Genomic Map