siRNA Details
| Gene Symbol: RERGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286201.1 | NP_001273130.1 | 5 | 538 | ||
| NM_024730.3 | NP_079006.1 | 6 | 695 | |||
| NR_104413.1 | 5 | 435 | ||||
| GenBank mRNA | AK026308.1 | - | 6 | 621 | ||
| BC042888.1 | - | 6 | 682 | 5198817 | ||
| BX422875.2 | - | 5 | 486 | |||

RERGL
Genomic Map