siRNA Details
| Gene Symbol: ZNF665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024733.3 | NP_079009.3 | 4 | 505 | ||
| XM_005259266.4 | XP_005259323.1 | 4 | 539 | |||
| XM_011527325.2 | XP_011525627.1 | 4 | 610 | |||
| XM_011527327.2 | XP_011525629.1 | 5 | 914 | |||
| GenBank mRNA | AK291403.1 | - | 4 | 495 | ||
| AL832507.1 | - | 4 | 490 | |||

ZNF665
Genomic Map