siRNA Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282683.1 | NP_001269612.1 | 7 | 606 | ||
| NM_024735.4 | NP_079011.3 | 6 | 858 | |||
| GenBank mRNA | AF318348.1 | - | 5 | 1058 | ||
| AF428140.1 | - | 6 | 822 | |||
| AL117444.1 | - | 4 | 366 | |||
| AY736035.1 | - | 7 | 809 | |||
| BC012748.1 | - | 6 | 832 | 3958783 | ||
| BG718342.1 | - | 7 | 605 | |||

FBXO31
Genomic Map