siRNA Details
| Gene Symbol: CLMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024769.3 | NP_079045.1 | 2, 3 | 545 | ||
| XM_011542998.2 | XP_011541300.1 | 2, 3 | 513 | |||
| GenBank mRNA | AK026068.1 | - | 2, 3 | 521 | ||
| AK304843.1 | - | 2, 3 | 492 | |||
| AY326422.1 | - | 2, 3 | 369 | |||
| AY358340.1 | - | 2, 3 | 369 | |||
| BC009371.2 | - | 2, 3 | 191 | 4122648 | ||
| BE272517.1 | - | 2, 3 | 439 | |||
| CA395814.1 | - | 2, 3 | 458 | |||

CLMP
Genomic Map