siRNA Details
| Gene Symbol: NHEJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024782.2 | NP_079058.1 | 2 | 250 | ||
| GenBank mRNA | AJ972687.1 | - | 2 | 194 | ||
| AK022672.1 | - | 2 | 186 | |||
| BC012732.2 | - | 2 | 171 | 4026495 | ||
| BC030986.1 | - | 2 | 203 | 4076851 | ||
| CR457291.1 | - | 1 | 104 | |||
| DB124934.1 | - | 2 | 250 | |||

NHEJ1
Genomic Map