siRNA Details
| Gene Symbol: FAM124B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122779.1 | NP_001116251.1 | 1 | 548 | ||
| NM_024785.2 | NP_079061.2 | 1 | 548 | |||
| GenBank mRNA | AK026399.1 | - | 1 | 588 | ||
| AK075126.1 | - | 1 | 51 | |||
| BC025754.1 | - | 1 | 548 | 5202295 | ||

FAM124B
Genomic Map