siRNA Details
| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025318.1 | - | 3 | 357 | ||
| AK296616.1 | - | 3 | 233 | |||
| BC099716.1 | - | 3 | 348 | 40005934 | ||
| BC105634.1 | - | 3 | 348 | 40005931 | ||
| DA890426.1 | - | 3 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBAL3| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025318.1 | - | 3 | 357 | ||
| AK296616.1 | - | 3 | 233 | |||
| BC099716.1 | - | 3 | 348 | 40005934 | ||
| BC105634.1 | - | 3 | 348 | 40005931 | ||
| DA890426.1 | - | 3 | 360 | |||