siRNA Details
| Gene Symbol: RBFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171967.1 | NP_001165438.1 | 3 | 394 | ||
| NM_024805.2 | NP_079081.2 | 3 | 394 | |||
| GenBank mRNA | AF261073.2 | - | 3 | 274 | ||
| AF439745.2 | - | 3 | 274 | |||
| AK024825.1 | - | 3 | 382 | |||
| AK124595.1 | - | 1 | 2268 | |||
| BC014195.2 | - | 3 | 256 | 3938406 | ||
| BC057772.1 | - | 3 | 262 | 30337652 | ||

RBFA
Genomic Map