siRNA Details
| Gene Symbol: PTCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284403.1 | NP_001271332.1 | 7 | 1176 | ||
| NM_001284404.1 | NP_001271333.1 | 8 | 1306 | |||
| NM_001284405.1 | NP_001271334.1 | 9 | 1424 | |||
| NM_024754.4 | NP_079030.3 | 10 | 1503 | |||
| XM_005248601.2 | XP_005248658.1 | 9 | 1383 | |||
| XM_005248603.2 | XP_005248660.1 | 9 | 1359 | |||
| XM_017009866.1 | XP_016865355.1 | 9 | 1364 | |||
| XM_017009867.1 | XP_016865356.1 | 8 | 1276 | |||
| XM_017009868.1 | XP_016865357.1 | 8 | 1245 | |||
| GenBank mRNA | AK022660.1 | - | 8 | 1265 | ||
| AK056761.1 | - | 10 | 2167 | |||
| AK298750.1 | - | 7 | 1167 | |||
| AK303639.1 | - | 8 | 1275 | |||
| BC018720.1 | - | 10 | 1479 | 4552964 | ||

PTCD2
Genomic Map