siRNA Details
| Gene Symbol: KDM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145348.1 | NP_001138820.1 | 8 | 2441 | ||
| NM_024773.2 | NP_079049.2 | 8 | 2391 | |||
| XM_017023676.1 | XP_016879165.1 | 7 | 2298 | |||
| XM_017023677.1 | XP_016879166.1 | 9 | 2476 | |||
| XM_017023678.1 | XP_016879167.1 | 9 | 2472 | |||
| XM_017023679.1 | XP_016879168.1 | 9 | 2472 | |||
| XM_017023680.1 | XP_016879169.1 | 6 | 1983 | |||
| GenBank mRNA | AK023860.1 | - | 8 | 2382 | ||
| BC027911.1 | - | 8 | 2370 | 5207043 | ||

KDM8
Genomic Map