siRNA Details
| Gene Symbol: FAM124B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122779.1 | NP_001116251.1 | 2 | 1560 | ||
| NM_024785.2 | NP_079061.2 | 3 | 1685 | |||
| GenBank mRNA | AK026399.1 | - | 2 | 1599 | ||
| AK075126.1 | - | 3 | 1188 | |||
| BC025754.1 | - | 3 | 1685 | 5202295 | ||

FAM124B
Genomic Map