siRNA Details
| Gene Symbol: RNF122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024787.3 | NP_079063.2 | 6 | 1412 | ||
| XM_017013846.1 | XP_016869335.1 | 6 | 1308 | |||
| GenBank mRNA | AK022588.1 | - | 6 | 1409 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF122| Gene Symbol: RNF122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024787.3 | NP_079063.2 | 6 | 1412 | ||
| XM_017013846.1 | XP_016869335.1 | 6 | 1308 | |||
| GenBank mRNA | AK022588.1 | - | 6 | 1409 | ||