siRNA Details
| Gene Symbol: KLF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206.2 | NP_001197.1 | 2 | 2078 | ||
| GenBank mRNA | BC069431.1 | - | 2 | 857 | 7262239 | |
| D31716.1 | - | 2 | 2048 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLF9| Gene Symbol: KLF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206.2 | NP_001197.1 | 2 | 2078 | ||
| GenBank mRNA | BC069431.1 | - | 2 | 857 | 7262239 | |
| D31716.1 | - | 2 | 2048 | |||