siRNA Details
| Gene Symbol: DCAKD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128631.2 | NP_001122103.1 | 5 | 1393 | ||
| NM_001288654.2 | NP_001275583.1 | 5 | 1529 | |||
| NM_001288655.1 | NP_001275584.1 | 5 | 1441 | |||
| NM_001321326.1 | NP_001308255.1 | 5 | 1339 | |||
| NM_024819.6 | NP_079095.3 | 5 | 1526 | |||
| XM_011525262.2 | XP_011523564.1 | 5 | 1558 | |||
| XM_017025105.1 | XP_016880594.1 | 6 | 1631 | |||
| XM_017025106.1 | XP_016880595.1 | 6 | 1573 | |||
| GenBank mRNA | BC006472.1 | - | 5 | 1405 | 2822306 | |
| BC006546.2 | - | 5 | 1405 | 2822306 | ||
| BC018132.2 | - | 4 | 1277 | 3903964 | ||
| BC033299.1 | - | 4 | 1416 | 4153431 | ||

DCAKD
Genomic Map