siRNA Details
| Gene Symbol: CAAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167575.1 | NP_001161047.1 | 6 | 1992 | ||
| NM_024828.3 | NP_079104.3 | 6 | 1838 | |||
| GenBank mRNA | AF131768.1 | - | 1 | 971 | ||
| AK023719.1 | - | 6 | 1812 | |||
| BC014658.1 | - | 6 | 1814 | 4648932 | ||
| BC071953.1 | - | 6 | 1737 | 5431506 | ||

CAAP1
Genomic Map