siRNA Details
| Gene Symbol: WDR76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167941.1 | NP_001161413.1 | 2 | 292 | ||
| NM_024908.3 | NP_079184.2 | 2 | 402 | |||
| XM_005254680.3 | XP_005254737.1 | 2 | 365 | |||
| GenBank mRNA | AK023035.1 | - | 2 | 307 | ||
| AK290933.1 | - | 2 | 324 | |||
| BC025247.1 | - | 2 | 333 | 4560352 | ||
| BC051855.1 | - | 2 | 305 | 6141090 | ||
| BX374706.2 | - | 2 | 218 | |||
| CN259143.1 | - | 2 | 343 | |||
| DA439086.1 | - | 2 | 402 | |||
| EF011624.1 | - | 2 | 305 | |||

WDR76
Genomic Map