siRNA Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031721.3 | NP_001026891.2 | 6 | 819 | ||
| NM_024840.3 | NP_079116.2 | 6 | 711 | |||
| XM_005259269.3 | XP_005259326.1 | 5 | 925 | |||
| XM_011527333.2 | XP_011525635.1 | 6 | 1035 | |||
| XM_017027315.1 | XP_016882804.1 | 5 | 883 | |||
| GenBank mRNA | AK027565.1 | - | 6 | 711 | ||
| BC057776.1 | - | 6 | 778 | 30338140 | ||

ZNF613
Genomic Map