siRNA Details
| Gene Symbol: DSN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145315.1 | NP_001138787.1 | 5 | 660 | ||
| NM_001145316.1 | NP_001138788.1 | 5 | 852 | |||
| NM_001145317.1 | NP_001138789.1 | 4 | 339 | |||
| NM_001145318.1 | NP_001138790.1 | 4 | 611 | |||
| NM_024918.3 | NP_079194.3 | 5 | 765 | |||
| XM_006723876.2 | XP_006723939.1 | 4 | 334 | |||
| XM_017028070.1 | XP_016883559.1 | 5 | 648 | |||
| XM_017028071.1 | XP_016883560.1 | 5 | 870 | |||
| XM_017028072.1 | XP_016883561.1 | 4 | 558 | |||
| GenBank mRNA | AK023408.1 | - | 6 | 491 | ||
| AK093031.1 | - | 5 | 760 | |||
| AK290738.1 | - | 5 | 852 | |||
| AK301671.1 | - | 4 | 544 | |||
| AK301840.1 | - | 4 | 289 | |||
| BC026011.1 | - | 4 | 865 | 4827570 | ||
| BC058899.1 | - | 5 | 618 | 6063417 | ||

DSN1
Genomic Map