siRNA Details
| Gene Symbol: FAM106CP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026810.1 | 1 | 325 | |||
| GenBank mRNA | BC098294.1 | - | 1 | 189 | ||
| BC098363.1 | - | 1 | 189 | |||
| DN989334.1 | - | 1 | 334 | |||
| Gene Symbol: FAM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026809.1 | 1 | 210 | |||
| GenBank mRNA | AK021862.1 | - | 1 | 210 | ||
| BC098263.1 | - | 1 | 189 | |||
| Gene Symbol: FAM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126160.1 | 1 | 325 | |||

FAM106CP
Genomic Map