siRNA Details
| Gene Symbol: C3orf36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025041.2 | NP_079317.2 | 1 | 1178 | ||
| GenBank mRNA | AK025826.1 | - | 1 | 1040 | ||
| AK225383.1 | - | 1 | 1040 | |||
| BC051274.1 | - | 1 | 1178 | |||
| BC064502.1 | - | 1 | 1178 | |||
| BC082257.1 | - | 1 | 1178 | |||
| BC104162.1 | - | 1 | 354 | 40029562 | ||

C3orf36
Genomic Map