siRNA Details
| Gene Symbol: LRRC8E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001268284.2 | NP_001255213.1 | 4 | 1713 | ||
| NM_001268285.2 | NP_001255214.1 | 2 | 1427 | |||
| NM_025061.5 | NP_079337.2 | 3 | 1570 | |||
| XM_011528319.2 | XP_011526621.1 | 3 | 2298 | |||
| GenBank mRNA | AK027073.1 | - | 1 | 518 | ||
| AK091134.1 | - | 3 | 1570 | |||
| AL834474.1 | - | 1 | 949 | |||
| BC022216.1 | - | 1 | 858 | |||
| BC070089.1 | - | 3 | 1570 | 30346745 | ||
| BC108252.1 | - | 1 | 679 | |||
| BX538180.1 | - | 3 | 1560 | |||

LRRC8E
Genomic Map