siRNA Details
| Gene Symbol: CCDC170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025059.3 | NP_079335.2 | 3 | 415 | ||
| XM_011536147.2 | XP_011534449.1 | 3 | 453 | |||
| XM_011536148.2 | XP_011534450.1 | 3 | 456 | |||
| GenBank mRNA | AK026958.1 | - | 3 | 328 | ||
| AK225330.1 | - | 3 | 329 | |||
| BC060803.1 | - | 3 | 364 | 5298914 | ||
| BI562667.1 | - | 3 | 369 | |||
| DB232703.1 | - | 3 | 415 | |||

CCDC170
Genomic Map