siRNA Details
| Gene Symbol: ZBTB48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278647.1 | NP_001265576.1 | 2 | 575 | ||
| NM_001278648.1 | NP_001265577.1 | 2 | 596 | |||
| NM_005341.3 | NP_005332.1 | 2 | 662 | |||
| XM_017001110.1 | XP_016856599.1 | 2 | 647 | |||
| XM_017001111.1 | XP_016856600.1 | 2 | 647 | |||
| XM_017001112.1 | XP_016856601.1 | 2 | 652 | |||
| XM_017001113.1 | XP_016856602.1 | 2 | 593 | |||
| XM_017001114.1 | XP_016856603.1 | 2 | 580 | |||
| GenBank mRNA | AK290156.1 | - | 2 | 570 | ||
| AK312786.1 | - | 2 | 578 | |||
| BC013573.2 | - | 1 | 521 | 3906038 | ||
| L16896.1 | - | 2 | 602 | |||

ZBTB48
Genomic Map