siRNA Details
| Gene Symbol: BBOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025057.2 | NP_079333.2 | 12 | 1982 | ||
| XM_011537171.2 | XP_011535473.1 | 13 | 2103 | |||
| XM_011537174.2 | XP_011535476.1 | 13 | 2096 | |||
| XM_017021660.1 | XP_016877149.1 | 11 | 1711 | |||
| XM_017021661.1 | XP_016877150.1 | 11 | 1756 | |||
| XM_017021662.1 | XP_016877151.1 | 10 | 1540 | |||
| XM_017021663.1 | XP_016877152.1 | 10 | 1349 | |||
| GenBank mRNA | AK026842.1 | - | 5 | 1015 | ||
| AL834487.1 | - | 7 | 1257 | |||

BBOF1
Genomic Map