siRNA Details
Gene Symbol: USP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321291.1 | NP_001308220.1 | 19, 20 | 3544 | ||
XM_005257542.2 | XP_005257599.1 | 19, 20 | 3568 | |||
XM_005257543.2 | XP_005257600.1 | 19, 20 | 3568 | |||
XM_011525065.1 | XP_011523367.1 | 19, 20 | 3582 | |||
XM_011525066.2 | XP_011523368.1 | 19, 20 | 3444 | |||
XM_011525067.2 | XP_011523369.1 | 19, 20 | 3444 | |||
XM_011525068.1 | XP_011523370.1 | 19, 20 | 3485 | |||
XM_011525069.2 | XP_011523371.1 | 18, 19 | 2847 | |||
XM_011525070.1 | XP_011523372.1 | 20, 21 | 3515 | |||
XM_017024900.1 | XP_016880389.1 | 19, 20 | 3568 | |||
XM_017024901.1 | XP_016880390.1 | 17, 18 | 3106 | |||
GenBank mRNA | AB040886.1 | - | 19, 20 | 3455 | ||
AK022840.1 | - | 14, 15 | 3508 | |||
AK022913.1 | - | 20, 21 | 3338 | |||
AK023077.1 | - | 1, 2 | 80 | |||
AK225155.1 | - | 2807 | ||||
AL833835.1 | - | 6, 7 | 1764 | |||
BC013890.2 | - | 4, 5 | 442 | |||
BC071582.1 | - | 19, 20 | 3544 | 30345255 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
