siRNA Details
| Gene Symbol: MAPK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002754.4 | NP_002745.1 | 12 | 1247 | ||
| NR_072996.1 | 10 | 1095 | ||||
| GenBank mRNA | AB209586.1 | - | 9 | 2053 | ||
| AF004709.1 | - | 12 | 1196 | |||
| AF015256.1 | - | 12 | 1208 | |||
| AF092535.1 | - | 12 | 1169 | |||
| AK222922.1 | - | 10 | 974 | |||
| BC000433.2 | - | 12 | 1189 | 2819932 | ||
| BC001641.2 | - | 12 | 1189 | 2819932 | ||
| BC004428.1 | - | 12 | 1189 | 2819932 | ||
| BC085196.1 | - | 10 | 1065 | 6107307 | ||
| U93232.1 | - | 12 | 1155 | |||
| Y10488.1 | - | 12 | 1170 | |||

MAPK13
Genomic Map