siRNA Details
| Gene Symbol: TRABD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320484.1 | NP_001307413.1 | 8 | 821 | ||
| NM_001320485.1 | NP_001307414.1 | 8 | 809 | |||
| NM_001320487.1 | NP_001307416.1 | 8 | 843 | |||
| NM_001320488.1 | NP_001307417.1 | 8 | 843 | |||
| NM_025204.3 | NP_079480.2 | 8 | 834 | |||
| NR_135275.1 | 8 | 805 | ||||
| XM_006724422.3 | XP_006724485.1 | 8 | 813 | |||
| XM_006724423.3 | XP_006724486.1 | 8 | 806 | |||
| XM_011530712.2 | XP_011529014.2 | 8 | 881 | |||
| XM_011530713.2 | XP_011529015.2 | 8 | 879 | |||
| XM_011530715.2 | XP_011529017.2 | 8 | 884 | |||
| XM_011530716.2 | XP_011529018.1 | 8 | 844 | |||
| XM_011530717.2 | XP_011529019.1 | 8 | 825 | |||
| XM_011530718.2 | XP_011529020.1 | 8 | 804 | |||
| XM_017028943.1 | XP_016884432.1 | 8 | 886 | |||
| XM_017028944.1 | XP_016884433.1 | 8 | 921 | |||
| GenBank mRNA | AF218001.1 | - | 1 | 969 | ||
| AK024447.1 | - | 1 | 3753 | |||
| AK292050.1 | - | 8 | 813 | |||
| AL449244.1 | - | 8 | 815 | |||
| AY203945.1 | - | 8 | 801 | |||
| BC012445.1 | - | 8 | 653 | 3857552 | ||
| BC093029.1 | - | 8 | 828 | 30527560 | ||
| BX389277.2 | - | 2 | 101 | |||
| BX417817.2 | - | 8 | 794 | |||
| BX425224.2 | - | 8 | 788 | |||

TRABD
Genomic Map